Labconco FreeZone Freeze Drying Chamber, Clear, Short; BT FD
Representative image only.
Labconco FreeZone Freeze Drying Chamber, Clear, Short; BT FD
Labconco | Mfr: 7318702| Item: 0335316
Easily attach flasks to your freeze dry system or contain samples in bulk
$4,420.17CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$4,420.17CAD / Each
Buy It Together
Current Item
Labconco FreeZone Freeze Drying Chamber, Clear, Short; BT FD
Labconco FreeZone Freeze Drying Chamber, Clear, Short; BT FD
0335316
$4,420.17CAD / Each
+ + + + =
Total Price 20587.68 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Clear acrylic lid, gasket, vacuum release valve, and pass through for electrical cords.
Key Features
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Obtain visibility while freeze drying samples in bulk
  • Accommodates short product shelves
  • Compatible with all FreeZone Freeze Dryers
  • 1 year warranty
More About this Item
These drying chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk. Various models are available from 8-, 12- to 16-port drying chambers to 3 to 5 heated product shelf chambers.
The short acrylic drying chamber is ideal for freeze drying numerous samples in bulk. It features an 8” diameter bottom opening to the collector and fits a 10-1/8” or 14” diameter condenser.
1 Year Warranty
Specifications
  • Exterior Height (InchShort)16.875
  • DescriptionFreeze Drying Chamber, Clear, Short; BT FD