Jenway 3510 pH Meter with FREE Electrode, 120 VAC

Representative image only.

Jenway 3510 pH Meter with FREE Electrode, 120 VAC

Jenway | Item: 3545913

Compact unit provides accurate and reliable readings

$1,480.76CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,480.76CAD / Each

Buy It Together

Current Item

Jenway 3510 pH Meter with FREE Electrode, 120 VAC
Jenway 3510 pH Meter with FREE Electrode, 120 VAC
3545913

$1,480.76CAD / Each
+ + =
Total Price 1943.77 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

FREE pH electrode and power cord.

Key Features

  • Supports one, two, or three point pH calibration
  • Up to three decimal place resolution for accurate readings
  • FREE pH electrode included
  • 3 year warranty

More About this Item

Benchtop pH meters feature up to three decimal place resolution, support up to three pH calibration points, and offer manual and automatic buffer recognition for DIN, JIS, and NIST buffers. A large LCD displays status and results of pH and temperature readings. Easily export data to a computer or printer via an RS-232 or an analog interface.

3 Year Warranty
Specifications
  • Meter TypeBenchtop
  • Display typeBacklit custom LCD
  • Min pH (pH)2
  • Max pH (pH)19.999
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.003 pH
  • Buffer RecognitionDIN, NIST, JIS, and custom
  • CalibrationUp to 3 points
  • Min mV (mV)-1999.9
  • Max mV (mV)1999.9
  • mV Resolution0.1/1
  • mV Accuracy±0.2 mV
  • Min Temperature (° C)10
  • Max Temperature (° C)105
  • Min Temperature (° F)14
  • Max Temperature (° F)221
  • Temperature Resolution1°F (0.1°C)
  • Temperature Accuracy±1°F (±0.5°C)
  • Temperature compensationAutomatic or manual from 32 to 212°F (0 to 100°C)
  • InterfaceRS-232
  • OutputAnalog and RS-232
  • Data Logging (points)32 data sets
  • Power (VAC)120
  • DescriptionpH Meter with FREE Tris Compatible Electrode, 120 VAC
  • Warranty3 year
Reviews
Q&A