Jenway 025 162 pH Buffer, 9.22 pH; 500 mL

Representative image only.

Jenway 025 162 pH Buffer, 9.22 pH; 500 mL

Jenway | Item: 7900032

All your pH measurement needs in one place

$78.83CAD / Each

$78.83CAD / Each

Buy It Together

Current Item

Jenway 025 162 pH Buffer, 9.22 pH; 500 mL
Jenway 025 162 pH Buffer, 9.22 pH; 500 mL
7900032

$78.83CAD / Each
+
Environmental Express Electrode Storage Solution; 500 mL
0065304

$40.22CAD / Pint of 1
+ + =
Total Price 402.97 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Jenway offers a range of pH buffers either as single use sachets or longer lasting 500 mL bottles

More About this Item

To care for your Jenway pH electrodes there is also a range of storage solutions and fill solutions tailored to your specific probe.

Specifications
  • Accessory TypepH Buffer
  • RangepH 9.22; 500 mL
  • DescriptionpH Buffer, 9.22 pH; 500 mL
Reviews
Q&A