Jenway 025 157 Redox Standard, 200 mV; 500 mL

Representative image only.

Jenway 025 157 Redox Standard, 200 mV; 500 mL

Jenway | Item: 7900029

All your pH measurement needs in one place

$252.32CAD / Each

$252.32CAD / Each

Buy It Together

Current Item

Jenway 025 157 Redox Standard, 200 mV; 500 mL
Jenway 025 157 Redox Standard, 200 mV; 500 mL
7900029

$252.32CAD / Each
+
Environmental Express Electrode Storage Solution; 500 mL
0065304

$40.22CAD / Pint of 1
+ + =
Total Price 576.46 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Jenway offers a range of pH buffers either as single use sachets or longer lasting 500 mL bottles

More About this Item

To care for your Jenway pH electrodes there is also a range of storage solutions and fill solutions tailored to your specific probe.

Specifications
  • Accessory TypeRedox Standard
  • Range200 mV; 500 mL
  • DescriptionRedox Standard, 200 mV; 500 mL
Reviews
Q&A