Idex Threaded Luer Adapter, Black PEEK, 0.050" Bore, Male Luer Lock x Female M6 Flat Bottom; 1/EA
Representative image only.
Idex Threaded Luer Adapter, Black PEEK, 0.050" Bore, Male Luer Lock x Female M6 Flat Bottom; 1/EA
Idex | Mfr: P-657| Item: 0202358
Convenience of quick connect and disconnect of a luer connection
$18.30CAD / Each
$25.64CAD / Each
$18.30CAD / Each
$25.64CAD / Each
Buy It Together
Current Item
Idex Threaded Luer Adapter, Black PEEK, 0.050" Bore, Male Luer Lock x Female M6 Flat Bottom; 1/EA
Idex Threaded Luer Adapter, Black PEEK, 0.050" Bore, Male Luer Lock x Female M6 Flat Bottom; 1/EA
0202358
$18.30CAD / Each
$25.64CAD / Each
+ + + + + + + + + =
Total Price 301.18 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Manufactured from inert polymers
  • Male Luer Lock x Female M6 Flat Bottom
  • Lock hub securely holds female luer adapters in place
  • 0.050" thru-hole diameter
  • Manufactured from PEEK
  • 1 year warranty
More About this Item
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
1 Year Warranty
Specifications
  • MaterialBlack PEEK
  • Connection Type-Port 1Male luer lock
  • Connection Type-Port 2Female M6 Flat Bottom
  • DescriptionThreaded Luer Adapter, Black PEEK, 0.050" Bore, Male Luer Lock x Female M6 Flat Bottom; 1/EA