Horiba 3200053514 Standard Nitrate 300 ppm, 14 mL bottles, 6/pk

Representative image only.

Horiba 3200053514 Standard Nitrate 300 ppm, 14 mL bottles, 6/pk

Horiba | Mfr: 3200053514| Item: 0576054

Optimize water analysis

$150.29CAD / Pkg of 6

$150.29CAD / Pkg of 6

Buy It Together

Current Item

Horiba 3200053514 Standard Nitrate 300 ppm, 14 mL bottles, 6/pk
Horiba 3200053514 Standard Nitrate 300 ppm, 14 mL bottles, 6/pk
0576054

$150.29CAD / Pkg of 6
+
Horiba U-52 Multiparameter Meter, 6-ft (2-M) Cable
9960022

$10,742.78CAD / Each
=
Total Price 10893.07 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose from a variety of accessories
  • 1 year warranty

More About this Item

Replacement sensors, probes, calibration standards, and other accessories for your Horiba meters.

1 Year Warranty
Specifications
  • DescriptionStandard Nitrate 300 ppm, 14 mL bottles, 6/pk
  • Warranty1 year
Documents & Certificates
Reviews
Q&A