Horiba U-52 Multiparameter Meter
Measure and display pH, mV, conductivity, TDS, salinity, dissolved oxygen, turbidity, seawater specific gravity, and temperature simultaneously
  • Auto-calibration feature simplifies calibration of pH, dissolved oxygen, conductivity, and turbidity
  • Rugged housing is both waterproof and shock resistant
  • Store up to 10,000 readings with USB connectivity for export of data to a PC
These handheld, waterproof meters feature a large 3-1/2" (8.9 cm), easy-to-read LCD with backlighting for use in dark and low-light conditions. Multiple parameters can be measured and displayed at the same time. USB/probe connectivity and battery power are also indicated via the display. Variable contrast allows you to adjust for extreme ambient light conditions.
Meters are designed for one-handed operation. Probes are attached via a quick-connect connector allowing for fast setup. Unique auto-calibration feature provides hassle-free meter setup. Turbidity sensor offers better accuracy than most other conventional water quality meters. Polarographic DO sensor features improved stability, fast response, and easy maintenance.
Auto-hold function freezes average data values providing more time to verify or transcribe data. Up to 10,000 data sets can be stored and later transferred to a PC via a built-in USB connection. Units of measure are user selectable which helps you save time.
$10,742.78 - $12,231.05CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Horiba U-52 Multiparameter Meter, 6-ft (2-m) Cable 9960022 Mfr # 3200164501
COMPARE
Multiparameter Meter, 6-ft (2-m) Cable
$10,742.78CAD / Each
Horiba U-52 Multiparameter Meter, 33-ft (10-m) Cable 9960023 Mfr # 3200164502
COMPARE
Multiparameter Meter, 33-ft (10-m) Cable
$11,101.70CAD / Each
Horiba U-52 Multiparameter Meter, 100-ft (30-m) Cable 9960024 Mfr # 3200164503
COMPARE
Multiparameter Meter, 100-ft (30-m) Cable
$12,231.05CAD / Each