Hach TNT850 TNTplus® Spectrophotometer Chemistry, Lead, 0.1 to 2.0 mg/L; 25/PK
Representative image only.
Hach TNT850 TNTplus® Spectrophotometer Chemistry, Lead, 0.1 to 2.0 mg/L; 25/PK
Hach | Mfr: TNT850| Item: 9951539
Perform common methods with these pre-made reagent tests
$197.43CAD / Pkg of 25
$197.43CAD / Pkg of 25
Buy It Together
Current Item
Hach TNT850 TNTplus® Spectrophotometer Chemistry, Lead, 0.1 to 2.0 mg/L; 25/PK
Hach TNT850 TNTplus® Spectrophotometer Chemistry, Lead, 0.1 to 2.0 mg/L; 25/PK
9951539
$197.43CAD / Pkg of 25
+
Hach 2667200 Iron, Mid Range Color Disc Test Kit
0554565
$170.91CAD / Each
+
Hach DR3900 Benchtop Visible Spectrophotometer
9951500
$11,043.06CAD / Each
+ =
Total Price 11499.36 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measures: Lead, 0.1 to 2.0 mg/L
More About this Item
Dozens of options to cover most applications!
Specifications
  • MaterialLead, 0.1 to 2.0 mg/L
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, Lead, 0.1 to 2.0 mg/L; 25/PK