Hach 2667200 Iron, Mid Range Color Disc Test Kit
Representative image only.
Hach 2667200 Iron, Mid Range Color Disc Test Kit
Hach | Mfr: 2667200| Item: 0554565
Color wheel method provides accurate visual comparisons
$170.91CAD / Each
$170.91CAD / Each
Buy It Together
Current Item
Hach 2667200 Iron, Mid Range Color Disc Test Kit
Hach 2667200 Iron, Mid Range Color Disc Test Kit
0554565
$170.91CAD / Each
+ + =
Total Price 1332.16 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestIron, mid range
  • Measurement Points 0.2 to 10 ppm
  • Reagent Platform1,10 Phenanthroline
  • Tests Per Kit100
  • DescriptionIron, Mid Range Color Disc Test Kit