Hach TNT82206 TNTplus® Spectrophotometer Chemistry, High Range COD, 20 to 1500 mg/L; 150/PK
Representative image only.
Hach TNT82206 TNTplus® Spectrophotometer Chemistry, High Range COD, 20 to 1500 mg/L; 150/PK
Hach | Mfr: TNT82206| Item: 9951518
Perform common methods with these pre-made reagent tests
$747.01CAD / Pkg of 6
$747.01CAD / Pkg of 6
Buy It Together
Current Item
Hach TNT82206 TNTplus® Spectrophotometer Chemistry, High Range COD, 20 to 1500 mg/L; 150/PK
Hach TNT82206 TNTplus® Spectrophotometer Chemistry, High Range COD, 20 to 1500 mg/L; 150/PK
9951518
$747.01CAD / Pkg of 6
+
Hach 2667200 Iron, Mid Range Color Disc Test Kit
0554565
$170.91CAD / Each
+
Hach DR3900 Benchtop Visible Spectrophotometer
9951500
$11,043.06CAD / Each
+ =
Total Price 12048.94 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measures: High Range COD, 20 to 1500 mg/L Package: 6 x 25
More About this Item
Dozens of options to cover most applications!
Specifications
  • MaterialHigh Range COD, 20 to 1500 mg/L
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, High Range COD, 20 to 1500 mg/L; 150/PK