Hach DR 900 Multiparameter Colorimeter, Handheld

Representative image only.

Hach DR 900 Multiparameter Colorimeter, Handheld

Hach | Mfr: 9385100| Item: 9957310

Select from 90 preprogrammed tests for the most common parameters

$3,630.43CAD / Each

$3,630.43CAD / Each

Buy It Together

Current Item

Hach DR 900 Multiparameter Colorimeter, Handheld
Hach DR 900 Multiparameter Colorimeter, Handheld
9957310

$3,630.43CAD / Each
+ +
Hach 2635300 Speccheck Standards, DPD for Chlorine
9957496

$444.96CAD / Each
=
Total Price 4539.51 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

two 25-mL glass vials, two 10-mL plastic vials, COD vial adapter, USB cable, and four AA batteries.

Key Features

  • "Favorites" list for easy method selection
  • Intuitive user interface allows quick selection and easy testing in 22 languages
  • Easy USB data transfer for up to 500 data sets
  • Rugged, waterproof housing is IP67 rated and permits on-site testing
  • 1 year warranty

More About this Item

The handheld DR 900 colorimeter allows quick and easy access to the most common water testing methods. Intuitive userinterface provides helpfulprompts in user-selectable languages. "Favorite" list saves your most commonly used methods for quick access.

Colorimeter features automatic wavelength selection, advanced reagent blank correction, and calibration curve adjustment. Analysis can be performed with user-set or preset reaction times with built-in timer. Large, backlit display reads results as concentration, absorbance, or percent transmittance.

Store up to 500 data sets with sample ID and user ID. Easy USB data transfer with no software or accessories needed; data simply transfers as .cvs file.

Durable over-molded housing is waterproof, dustproof, and shockproof; ideal for demanding field environments.

1 Year WarrantyConformite EuropeenneEPA United States Environmental protection agencyIP 67
Specifications
  • Display TypeLED
  • Wavelength(s) (nm)420, 520, 560, or 610
  • Min Absorbance0
  • Max Absorbance2
  • Test Parameters90 preprogrammed methods
  • WaterproofYes
  • Data LoggingYes
  • Memory Size500 measurements
  • OutputUSB
  • Battery4 AA (included)
  • Length (in)9 1/8
  • Width (in)3 13/16
  • Height (in)1 7/8
  • DescriptionMultiparameter Colorimeter, Handheld
  • Warranty1 year
Reviews
Q&A