Hach DR 300 600 nm Pocket Colorimeter

Representative image only.

Hach DR 300 600 nm Pocket Colorimeter

Hach | Mfr: LPV445.97.60110| Item: 9957392

Single-parameter colorimeter you can take anywhere

$1,047.58CAD / Each

$1,047.58CAD / Each

Buy It Together

Current Item

Hach DR 300 600 nm Pocket Colorimeter
Hach DR 300 600 nm Pocket Colorimeter
9957392

$1,047.58CAD / Each
+ +
Environmental Express Model 200 COD Colorimeter
9956184

$1,492.54CAD / Each
+ + +
Environmental Express Model 200 Chlorine Colorimeter
9956178

$1,280.38CAD / Each
+ =
Total Price 4261.12 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

Includes

carrying case, four AAA alkaline batteries, sample cells, and Hach Communication Dongle. Reagents are sold separately.

Key Features

  • Rugged design withstands water, drops, extreme temperatures, and dirt
  • Compact size fits easily into a kit or your pocket
  • Read the large digital display in any condition—features backlight
  • No need to calibrate—just blank and read!
  • Store and recall up to 50 measurements
  • 1 year warranty

More About this Item

The DR300 series continues Hach's legacy of single-parameter colorimeters for water testing. These units are factory calibrated, so you only need to zero the unit before taking readings. Secondary calibration sets are available for some parameters to meet regulatory requirements. New features include a more intuitive interface to reduce potential manual errors, and larger digits on the backlit display.

Range will depend upon test used.

1 Year Warranty
Specifications
  • Display TypeLCD
  • Light SourceLED
  • Detector TypeSilicon Photodiode
  • Wavelength(s) (nm)Fixed
  • Min Absorbance0
  • Max Absorbance2.5
  • Absorbance Accuracy±2 nm
  • Test Parameters600 nm
  • Min Operating Temperature (° F)32
  • Max Operating Temperature (° F)122
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° C)50
  • WaterproofYes
  • Data Logging50 measurements
  • Battery4 AAA (included)
  • Length (cm)15.7
  • Width (cm)6.9
  • Height (cm)3.4
  • Description600 nm Pocket Colorimeter
  • Warranty1 year
Reviews
Q&A