Hach 223102 Free Chlorine Color Disc Test Kit , Range 0 to 3.4 ppm
Representative image only.
Hach 223102 Free Chlorine Color Disc Test Kit , Range 0 to 3.4 ppm
Hach | Mfr: 223102| Item: UZ-05545-74
Color wheel method provides accurate visual comparisons
$142.85CAD / Each
$142.85CAD / Each
Buy It Together
Current Item
Hach 223102 Free Chlorine Color Disc Test Kit , Range 0 to 3.4 ppm
Hach 223102 Free Chlorine Color Disc Test Kit , Range 0 to 3.4 ppm
0554574
$142.85CAD / Each
+ + =
Total Price 1304.10 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestChlorine, free
  • Measurement Points0 to 3.4 ppm
  • Reagent PlatformDPD Method
  • Tests Per Kit100
  • DescriptionFree Chlorine Color Disc Test Kit , Range 0 to 3.4 ppm