Global Water SP200 Variable Speed, Peristaltic, Fluid Sampling Pump
Representative image only.
Global Water SP200 Variable Speed, Peristaltic, Fluid Sampling Pump
Global Water | Mfr: CJ0000| Item: 9924305
Ideal for sampling lakes, ponds, holding pools, or shallow wells
$2,118.73CAD / Each
$2,118.73CAD / Each
Buy It Together
Current Item
Global Water SP200 Variable Speed, Peristaltic, Fluid Sampling Pump
Global Water SP200 Variable Speed, Peristaltic, Fluid Sampling Pump
9924305
$2,118.73CAD / Each
+
Manning PSB4A2A2A1F2 Portable Peristaltic Sampler
9924000
$4,724.72CAD / Each
+ +
Global Water WS700 Peristaltic Water Sampler
9924370
$4,482.05CAD / Each
+ =
Total Price 15193.14 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Simple control for exact sample size
  • Reversible motors for backflushing of sample hose
  • Rugged, UV-protected PVC case with handle for easy use and transportation
  • Flow rate : up to 500 mL/min at 4-ft head, operating temperature: 0 to 70C
  • Features a 10 ft (3.05 m) long power cord fitted with alligator clips for easy connection
  • 1 year warranty
More About this Item
These portable sampling pumps are ideal for sampling lakes, ponds, holding pools, or shallow wells. Samplers are packaged in a lightweight, rugged, UV protected PVC case with handle for easy use and transportation. To protect sample integrity the sample only comes in contact with Norprene and polyethylene tubing which is easily cleaned or replaced between samples.
1 Year Warranty
Specifications
  • Sampling PumpPeristaltic
  • TubingReinforced PVC (1/4" ID)
  • Power12 VDC battery (not included)
  • DescriptionVariable Speed, Peristaltic, Fluid Sampling Pump