Fowler 54422444 Digi-Level Electronic Protractor, 0 to 360 ° (4 x 90 °) range
Representative image only.
Fowler 54422444 Digi-Level Electronic Protractor, 0 to 360 ° (4 x 90 °) range
Fowler | Mfr: 544224440| Item: 0991871
Complete 360° angle measurements
$512.37CAD / Each
for an additional
$162.81CAD / Each
for an additional
$288.78CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$512.37CAD / Each
Buy It Together
Current Item
Fowler 54422444 Digi-Level Electronic Protractor, 0 to 360 ° (4 x 90 °) range
Fowler 54422444 Digi-Level Electronic Protractor, 0 to 360 ° (4 x 90 °) range
0991871
$512.37CAD / Each
+ =
Total Price 675.18 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
A digital LCD for inclination values. Measurements are generated by an electronic gravity sensor and processed through an electronic circuit. Other features include alternate zero (reference), display hold, and a machined aluminum frame.
1 Year Warranty
Specifications
  • Min range (degrees)0
  • Max range (degrees)360
  • Resolution0.05°
  • DescriptionDigi-Level Electronic Protractor, 0 to 360 ° (4 x 90 °) range