Fluke TLK-220 SureGrip™ Industrial Test Lead Kit with Case
Representative image only.
Fluke TLK-220 SureGrip™ Industrial Test Lead Kit with Case
Fluke | Mfr: TLK-220| Item: UZ-20037-35
High performance at an economical price
$343.30CAD / Each
$343.30CAD / Each
Buy It Together
Current Item
Fluke TLK-220 SureGrip™ Industrial Test Lead Kit with Case
Fluke TLK-220 SureGrip™ Industrial Test Lead Kit with Case
2003735
$343.30CAD / Each
+ =
Total Price 704.28 ADD 2 ITEMS TO CART
Product Description
Includes
AC220 plunger style alligator clips, AC285 large jaw alligator clips, TP220 sharp test probes, TL224 right to straight test leads, and vinyl carrying case.
Key Features
  • SureGrip™ clips, probes, and leads in a convenient carrying case
  • 1000 V CAT III, 600 V CAT IV rated
  • 1 year warranty
More About this Item
Various test leads are available to suit your application.
SureGrip™ set is ideal for industrial applications. All CAT III-1000 V, CAT IV-600 V rated.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • DescriptionSureGrip™ Industrial Test Lead Kit with Case