Fluke TLK-225 SureGrip™ Master Accessory Kit with Pouch
Representative image only.
Fluke TLK-225 SureGrip™ Master Accessory Kit with Pouch
Fluke | Mfr: TLK-225| Item: 2000854
High performance at an economical price
$371.28CAD / Each
$371.28CAD / Each
Product Description
Includes
TP220 probes, TL224 silicones leads, AC280 hooks, AC283 grabbers, both AC220 and AC285 small and large alligator clips, and a pouch.
Key Features
  • Ideal replacement kit for all SureGrip™ leads and probes
  • CAT II 1000 V, CAT III 1000 V, CAT IV 600 V rated
  • 1 year warranty
More About this Item
Various test leads are available to suit your application.
Complete set of premium SureGrip™ test accesssories with roll-up, 6-pocket storage pouch. Modular set design provides ultimater flexibility, with TP220 probes, TL224 silicones leads, AC280 hooks, AC283 grabbers, and both AC220 and AC285 small and large alligator clips. Rated CAT lll 1000 V, CAT lV 600 V, 10 A.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • Safety RatingCAT II 1000 V, CAT III 1000 V, CAT IV 600 V
  • DescriptionSureGrip™ Master Accessory Kit with Pouch