Fluke TL27 Heavy-Duty Test Lead Set
Representative image only.
Fluke TL27 Heavy-Duty Test Lead Set
Fluke | Mfr: TL27| Item: 2000848
High performance at an economical price
$85.46CAD / Each
$85.46CAD / Each
Buy It Together
Current Item
Fluke TL27 Heavy-Duty Test Lead Set
Fluke TL27 Heavy-Duty Test Lead Set
2000848
$85.46CAD / Each
+ =
Total Price 446.44 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Heavy-duty EPDM insulation offer excellent protection
  • One pair (red and black) DMM test leads with safety shrouded, standard diameter banana plugs
  • 1.5 m (60") long
  • CAT III 1000 V, 10 A rated
  • 1 year warranty
More About this Item
Various test leads are available to suit your application.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • Safety RatingCAT III 1000 V, 10 A
  • DescriptionHeavy-Duty Test Lead Set