Fluke 83-5 High Accuracy Digital Multimeter
Representative image only.
Fluke 83-5 High Accuracy Digital Multimeter
Fluke | Mfr: FLUKE-83-5| Item: 2608302
Durable, rugged design is ideal for industrial environments
$795.62CAD / Each
for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$795.62CAD / Each
Buy It Together
Current Item
Fluke 83-5 High Accuracy Digital Multimeter
Fluke 83-5 High Accuracy Digital Multimeter
2608302
$795.62CAD / Each
+ + =
Total Price 1321.23 ADD 3 ITEMS TO CART
Product Description
Includes
TL75 test leads, AC72 alligator clips, CD-ROM (user's manual and technical notes), operator's guide, holster, and 9 V battery (installed).
Key Features
  • Easily troubleshoot industrial equipment to prevent costly downtime
  • Large digit backlight LCD display is easy to read
  • Limited Lifetime Warranty
More About this Item
Available for sale in the United States and Canada only
The Fluke 83V industrial multimeter features a 6000-count, 3-3/4” digit display with a 32 segment bar graph. The 83V is CAT III 1000 V and CAT IV 600 V rated and can withstand 8000 volt spikes caused by load switching on industrial circuits. Meter is auto and manual ranging for maximum flexibility during testing. Meter provides Min/Max and average readings allowing you to capture variations.
UL Listed.Canadian Standards AssociationLimited Lifetime Warranty
Specifications
  • DC Max (Volts)1000
  • Current (AC/DC)10 A
  • AC Max (Volts)1000
  • Resistance Max (MΩ)60
  • Frequency Max (KHz)200
  • Count20000
  • Digits4.5
  • Conductance50
  • Safety RatingCAT III 1000V, CAT III 600V
  • DescriptionHigh Accuracy Digital Multimeter
  • WarrantyLTD Life Time