Fluke 725 US Multifunction Process Calibrator
Representative image only.
Fluke 725 US Multifunction Process Calibrator
Fluke | Mfr: FLUKE-725 US| Item: 2607250
Measure and source functions allow you to test and calibrate almost anything!
$6,381.46CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$6,381.46CAD / Each
Buy It Together
Current Item
Fluke 725   US Multifunction Process Calibrator
Fluke 725 US Multifunction Process Calibrator
2607250
$6,381.46CAD / Each
+ + =
Total Price 6978.90 ADD 3 ITEMS TO CART
Product Description
Includes
test leads (Fluke TL75), test clips, (Fluke AC72), one set of stackable test leads, NIST-traceable calibration report supplied by manufacturer, four AA batteries
Key Features
  • Easy to read measure/source screen allows you view input and output simultaneously
  • Perform rapid linearity tests with auto step and auto ramp features
  • Rugged, streamlined design is ideal for field conditions
  • 3 year warranty
More About this Item
Use this multifunction process calibrator to measure volts, mA, RTD’s, thermocouples, frequency, and ohms to test sensors and transmitters. The dual channel functionality of this unit allows you to source/simulate volts, mA, thermocouples, RTD’s, frequency, ohms, and pressure to easily calibrate transmitters.
Use this process calibrator to support flowmeter testing with the frequency and counts per minute functions provided. The frequently used function provides memory of test setups for future use and can store up to eight setups on any sourced function. Power your transmitters during test using the loop supply with simultaneous mA measurement.
This versatile process calibrator will measure/source pressure using any of the 29 Fluke 700Pxx pressure modules and has the ability to source mA with simultaneous pressure measurement to conduct valve and I/P tests. The rugged design is ideal for harsh field conditions and the simple pushbutton user interface is backlit for working in poor lighting conditions.
Available for sale in the United States and Canada only.
3 Year WarrantyCanadian Standards Association
Specifications
  • Pressure Resolution0.01 mV; 0.01 V (Source); 0.01 V (measure); 0.001 (mA); 0.01 to 0.1 Ω(Resistance); 2.0 to 1,000.0 CPM = 0.1 CPM (Frequency); 1 to 1100 Hz = 1 Hz (Frequency); 1.0 to 10.0 kHz = 0.1 kHz (Frequency)
  • Width (in)7 7/8
  • Height (in)1 7/8
  • Length (in)3 3/4
  • Batteries4 x AA batteries
  • DescriptionMultifunction Process Calibrator
  • Warranty3 Years