Fluke 17B+ CAT III Digital Multimeter with Temperature Measurement

Representative image only.

Fluke 17B+ CAT III Digital Multimeter with Temperature Measurement

Fluke | Mfr: FLUKE-17B+ ESP| Item: 1778514

Professional grade multimeter with temperature measurement function

$254.88CAD / Each

for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$254.88CAD / Each

Buy It Together

Current Item

Fluke 17B+ CAT III Digital Multimeter with Temperature Measurement
Fluke 17B+ CAT III Digital Multimeter with Temperature Measurement
1778514

$254.88CAD / Each
+ +
Fluke TPAK ToolPak Magnetic Meter Hanging Kit
2000856

$89.86CAD / Each
=
Total Price 540.33 ADD 3 ITEMS TO CART
Product Description

Includes

TL75 Hard Point™ test lead set, type K thermocouple probe, and batteries.

Key Features

  • Safety rated for CAT III 600 V
  • Take temperature measurements with included type K thermocouple probe
  • Measures AC/DC voltage up to 1000 V
  • Measures AC/DC current up to 10 A
  • Large, easy-to-read, backlit display
  • Features MIN/MAX and HOLD functions
  • Supports relative measurements to measure changes
  • Hazardous voltage alert LED help ensure operator safety
  • Easily operate even with gloves on
  • 1 year warranty

More About this Item

Compact, easy-to-use, and full-featured multimeter is a perfect tool for checking and diagnosing issues on electrical equipment, appliances, outlets, or switches. The meter measures AC/DC voltage, AC/DC current, resistance, frequency, duty cycle, and temperature as well as perform continuity and diode tests with excellent reliability and precision. Measure temperatures of refrigeration systems, heat exchangers, and line sets with the included type K thermocouple probe; select to view temperature in °C or °F.

Easily view measurements on a large 4000-count backlit LCD. Press the MIN/MAX or HOLD button to see minimum, maximum, or to freeze current reading. Use the relative measurement function to zero out the source reading to measure changes in voltage drop or temperature—ideal for identifying and diagnosing issues. Auto-shutoff feature turns off the meter after 20 minutes of inactivity, conserving battery life.

Conformite EuropeenneCanadian Standards Association1 Year Warranty
Specifications
  • DC Max (Volts)1000
  • DC Voltage Accuracy±(0.5% of reading + 3 digits)
  • DC Max (amps)10
  • DC Current Accuracy±(1.5% of reading + 3 digits)
  • AC Max (Volts)1000
  • Ac Voltage Accuracy±(1.0% of reading + 3 digits)
  • AC Max (amps)10
  • AC Current Accuracy±(1.5% of reading + 3 digits)
  • Resistance Max (MΩ)40
  • Capacitance Max (mF)1
  • Capacitance Accuracy40 and 400 nF ranges: ±(2% of reading + 5 digits)
    4, 40, 400, 1000 µF (1 mF) ranges: ±(5% of reading + 5 digits)
  • Frequency Max (KHz)100
  • Frequency Accuracy±(0.1% of reading + 3 digits)
  • Resistivity Accuracy400 Ω range: ±(0.5% of reading + 3 digits)
    4, 40, 400 kΩ, and 4 MΩ ranges: ±(0.5% of reading + 2 digits)
    40 MΩ range: ±(1.5% of reading + 3 digits)
  • ContinuityBeeper at ≤70 Ω
  • Diode Tester2 V
  • Duty Cycle1 % to 99 %
  • True RmsNo
  • Min Temperature (° F)-67
  • Max Temperature (° F)752
  • Min Temperature (° C)-55
  • Max Temperature (° C)400
  • Temperature Accuracy-55.0 to 0 °C: 9% +2 °C
    0 to 50.0 °C: 2 °C
    50.0 to 400.0 °C: 2% +1 °C
  • Count4000
  • DisplayLarge LCD
  • Safety RatingIEC 61010-1, IEC61010-2-030 CAT III 600 V, Pollution Degree 2
  • Power2 AA (included)
  • Width (in)3.5625
  • Height (in)7.25
  • Depth (in)2
  • Width (cm)9.1
  • Height (cm)18.3
  • Depth (cm)5
  • Description17B+ CAT III Digital Multimeter with Temperature Measurement
  • Warranty1 year
Reviews
Q&A