Fixed-Volume Lightweight Pipettor, Metal, Red, 1000 µL

Representative image only.

Fixed-Volume Lightweight Pipettor, Metal, Red, 1000 µL

| Item: 0783936

Compact size and durability makes pipettes ideal for field work

$375.35CAD / Each

for an additional
$155.44CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$375.35CAD / Each

Buy It Together

Current Item

Fixed-Volume Lightweight Pipettor, Metal, Red, 1000 µL
Fixed-Volume Lightweight Pipettor, Metal, Red, 1000 µL
0783936

$375.35CAD / Each
+ + =
Total Price 616.25 ADD 3 ITEMS TO CART
Product Description

Key Features

  • Highly accurate pipettors with durable, yet lightweight, metal construction
  • One-stroke plunger movement provides precise results
  • Fully Autoclavable to avoid cross-contamination
  • Color coded by volumes for easy identification
  • 1 year warranty

More About this Item

For accurate and consistent results, choose a fixed-volume pipettor. Ideal for repetitive delivery of reagents and solutions, these durable lightweight pipettors feature a precision-machined plunger and barrel assembly. The internal polyurethane multi-ring seals prevent fluid leakage far superior to single O-ring pipettors. Pipettors are color-coded to help you keep track of the proper dispensing volume — an excellent choice for a busy lab!

1 Year Warranty
Specifications
  • SterileNo
  • Max Volume (µL)1000
  • AutoclavableYes
  • Accuracy (%)±0.20%
  • Reproducibility±0.13%
  • Overall Length (InchShort)8
  • Overall Length (mm)203.2
  • Number Of Channels1
  • Tip EjectorNo
  • Body MaterialMetal
  • Color CodeRed
  • DescriptionFixed-Volume Lightweight Pipettor, Metal, Red, 1000 µL
  • Warranty1 year
Reviews
Q&A