Extech EX900 Waterproof Pocket Water Quality Tester

Representative image only.

Extech EX900 Waterproof Pocket Water Quality Tester

Extech | Mfr: EX900| Item: 9959825

Everything needed for pH, ORP, chlorine, and temperature measurement in one convenient kit

$499.48CAD / Each

for an additional
$153.97CAD / Each
for an additional
$276.26CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$499.48CAD / Each

Buy It Together

Current Item

Extech EX900 Waterproof Pocket Water Quality Tester
Extech EX900 Waterproof Pocket Water Quality Tester
9959825

$499.48CAD / Each
+ +
Extech RE305 Exstik Replacement ORP Electrode
3565003

$117.87CAD / Each
+
Extech EX800 Waterproof Pocket Water Quality Tester
9959820

$455.28CAD / Each
=
Total Price 1226.60 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

pocket tester; pH, ORP, and chlorine sensors; protective sensor cap; 50 chlorine reagent tablets; plastic sample cups; weighted stand; pH 4, 7, and 10 buffer pack; neck strap; carrying case; and four SR44 silver oxide button-cell batteries.

Key Features

  • Interchangeable flat surface electrodes work in liquids, semisolids, and solids
  • Smart chip informs you when to calibrate or replace your meter
  • Built-in memory recalls up to 15 labeled readings
  • 1 year warranty

More About this Item

Easy-to-use water quality pocket tester kit features everything needed for water testing and measurement. Reliable pocket tester with interchangeable sensors for pH, ORP, and chlorine allows you to be flexible in the field. Bundled calibration solutions and sample cups give you all the necessary accessories to begin taking readings.

Additional tester features include a built-in smart chip that simplifies calibration and error diagnosis. LCD simultaneously shows ppm or pH, and °F/°C temperature. An analog bar graph of measured values lets you view reading trends. Waterproof, IP57-rated tester floats in water.

1 Year Warranty
Specifications
  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • Min mV (mV)-1000
  • Max mV (mV)1000
  • mV Resolution1 mV
  • mV Accuracy±1mV + 1 digit
  • Min ISE0.01 to 10.00 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±10% of reading
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Min Temperature (° F)23
  • Max Temperature (° F)122
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 SR44 Silver Oxide (included)
  • Width (cm)3.4925
  • Height (cm)4.1275
  • Width (in)1 3/8
  • Length (in)6 15/16
  • Height (in)1 5/8
  • DescriptionWaterproof Pocket Water Quality Tester
  • Warranty1 year
Reviews
Q&A