Extech 42280 Humidity/Temperature USB Datalogger
Representative image only.
Extech 42280 Humidity/Temperature USB Datalogger
Extech | Mfr: 42280A| Item: 6851701
Datalogs up to 16,000 readings with USB interface for downloading to a PC
$440.54CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$440.54CAD / Each
Buy It Together
Current Item
Extech 42280 Humidity/Temperature USB Datalogger
Extech 42280 Humidity/Temperature USB Datalogger
6851701
$440.54CAD / Each
+ + +
Traceable Logger-Trac™ Temperature Data Logger
1800500
$175.33CAD / Each
+ =
Total Price 1760.84 ADD 5 ITEMS TO CART
Product Description
Includes
Windows® compatible software, USB cable, 110V AC adaptor and four AA batteries
Key Features
  • Triple LCD displays humidity, temperature and date (year, month and day)
  • Programmable from keypad or PC
  • Selectable data sampling rate
  • User programmable audible and visual alarms
  • Wall, desktop or tripod mount
  • Calibration via optional salt bottles
  • 1 year warranty
1 Year WarrantyConformite Europeenne
Specifications
  • Number of Channels1
  • Input TypeTemperature and humidity
  • Max Temperature (° F)144
  • Min Temperature (° F)-4
  • Max Temperature (° C)70
  • Min Temperature (° C)-20
  • Temperature Accuracy (° F)±1.0 (32 to 122), ±2.0 (all other ranges)
  • Temperature Accuracy (° C)±0.6 (0 to 50), ±1.2 (all other ranges)
  • Max Humidity RH (RH)100
  • Min Humidity RH (RH)0
  • Humidity Accuracy±3% from 10 to 90% (±5% outside of this range)
  • Memory Size16,000 total (8000 temperature and 8000 humidity readings)
  • Battery Type4 AA (included)
  • Display TypeMulti-function LCD with status LED indicators
  • DescriptionHumidity/Temperature USB Datalogger