Environmental Express Oakton pH Indicator Strips, 5 to 9, 100/pk
Representative image only.
Environmental Express Oakton pH Indicator Strips, 5 to 9, 100/pk
Vivid and easy-to-distinguish colors at every pH interval
$54.15CAD / Pkg of 100
$54.15CAD / Pkg of 100
Buy It Together
Current Item
Environmental Express Oakton pH Indicator Strips, 5 to 9, 100/pk
Environmental Express Oakton pH Indicator Strips, 5 to 9, 100/pk
3585004
$54.15CAD / Pkg of 100
+ + + + =
Total Price 531.61 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 3 month warranty
More About this Item
Convenient flip-top vials include color comparator chart. Read results immediately after exposing test area to the solution. Strips measure 2-3/4"L x 1/4"W (7 x 2 cm). Each vial contains 100 strips.
3 Month Warranty
Specifications
  • TestpH
  • Measurement Points5 to 9 pH in 0.5 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionpH Indicator Strips, 5 to 9, 100/pk