EMD Millipore 10020-1 Water Test Strips, Nitrate, 10 to 500 ppm, 100/Pk
Representative image only.
EMD Millipore 10020-1 Water Test Strips, Nitrate, 10 to 500 ppm, 100/Pk
EMD Millipore | Mfr: 1.10020.0001| Item: 9957213
Use for quick spot checks to ensure water quality
$151.32CAD / Each
$151.32CAD / Each
Buy It Together
Current Item
EMD Millipore 10020-1 Water Test Strips, Nitrate, 10 to 500 ppm, 100/Pk
EMD Millipore 10020-1 Water Test Strips, Nitrate, 10 to 500 ppm, 100/Pk
9957213
$151.32CAD / Each
+ + =
Total Price 285.62 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.
Specifications
  • TestNitrate
  • Measurement Points10 to 500 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionWater Test Strips, Nitrate, 10 to 500 ppm, 100/Pk