EMD Millipore 9590-3 PH Test Strips, 0-14 PH. Six Pks of 100.
Representative image only.
EMD Millipore 9590-3 PH Test Strips, 0-14 PH. Six Pks of 100.
EMD Millipore | Mfr: 1.09535.0007| Item: 9957208
Use for quick spot checks to ensure water quality
$275.23CAD / Pkg of 6
$275.23CAD / Pkg of 6
Buy It Together
Current Item
EMD Millipore 9590-3 PH Test Strips, 0-14 PH. Six Pks of 100.
EMD Millipore 9590-3 PH Test Strips, 0-14 PH. Six Pks of 100.
9957208
$275.23CAD / Pkg of 6
+ + =
Total Price 409.53 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.
The pH Test Strips are sold in six packs of 100.
Specifications
  • TestpH
  • Measurement Points0 to 14 pH in 1 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit600
  • DescriptionPH Test Strips, 0-14 PH. Six Pks of 100.