Watersafe Water Test Kits
Lab-quality results in a simple-to-use, affordable kit
  • Comprehensive test kits detect most common water contaminants
  • Compare your results against EPA-recommended levels
Fast, easy-to-use water test kits detect hazardous contaminants in municipal and private well drinking
water. Drinking water test kits indicate the presence of bacteria, pesticides, and lead. Kits also include readings for nitrate (0 to 50 ppm), nitrite (0 to 10 ppm), total chlorine (0 to 10 ppm), pH (6 to 10), and water hardness (0 to 425 ppm). Well water test kits add tests for contaminants commonly found in private well water: iron and copper (0 to 5 ppm). Order bacteria, pesticides, and lead stand-alone kits if less comprehensive tests are required. All kits are for one-time use.
$55.99 - $62.62CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Silver Lake Research WS-425B Drinking Water Test Kit , Single Use RK-99530-02 Mfr # WS-425B
COMPARE
Drinking Water Test Kit , Single Use
$62.62CAD / Each
Silver Lake Research WS-831 Bacteria Test Kit , Single Use RK-99530-06 Mfr # WS-831
COMPARE
Bacteria Test Kit , Single Use
$61.88CAD / Each
Silver Lake Research WS-207 Lead Test Kit , Single Use RK-99530-12 Mfr # WS-207
COMPARE
Lead Test Kit , Single Use
$55.99CAD / Each