EMD Millipore 10044-1 Water Test Strips, Cyanide, 0 to 30 ppm, 100/Pk
Representative image only.
EMD Millipore 10044-1 Water Test Strips, Cyanide, 0 to 30 ppm, 100/Pk
EMD Millipore | Mfr: 1.10044.0001| Item: 9957204
Use for quick spot checks to ensure water quality
$254.90CAD / Pkg of 100
$254.90CAD / Pkg of 100
Buy It Together
Current Item
EMD Millipore 10044-1 Water Test Strips, Cyanide, 0 to 30 ppm, 100/Pk
EMD Millipore 10044-1 Water Test Strips, Cyanide, 0 to 30 ppm, 100/Pk
9957204
$254.90CAD / Pkg of 100
+ + =
Total Price 389.20 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.
Kit includes reagent(s) to facilitate testing.
Specifications
  • TestCyanide
  • Measurement Points0, 1, 3, 10, 30 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionWater Test Strips, Cyanide, 0 to 30 ppm, 100/Pk