EMD Millipore 10025-1 Total Hardness Test Strips, 0 to 370 ppm
Representative image only.
EMD Millipore 10025-1 Total Hardness Test Strips, 0 to 370 ppm
EMD Millipore | Mfr: 1.10025.0001| Item: 9957210
Use for quick spot checks to ensure water quality
$184.03CAD / Pkg of 100
$184.03CAD / Pkg of 100
Buy It Together
Current Item
EMD Millipore 10025-1 Total Hardness Test Strips, 0 to 370 ppm
EMD Millipore 10025-1 Total Hardness Test Strips, 0 to 370 ppm
9957210
$184.03CAD / Pkg of 100
+ + =
Total Price 318.33 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.
Specifications
  • TestHardness
  • Measurement Points0 to 370 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionTotal Hardness Test Strips, 0 to 370 ppm