Davis Instruments 7747 Day-time Fan Aspirated Radiation Shield Kit
Representative image only.
Davis Instruments 7747 Day-time Fan Aspirated Radiation Shield Kit
Davis Instruments | Mfr: 7747| Item: 3000342
Available for sale in the United States and Canada only.
$548.84CAD / Each
$548.84CAD / Each
Buy It Together
Current Item
Davis Instruments 7747 Day-time Fan Aspirated Radiation Shield Kit
Davis Instruments 7747 Day-time Fan Aspirated Radiation Shield Kit
3000342
$548.84CAD / Each
+ =
Total Price 1601.65 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with Vantage Pro2 only; not compatible with original Vantage Pro.
  • 1 year warranty
More About this Item
Get the most from your Davis Instruments weather stations with these accessories and components.
Add the benefits of fan aspiration to your wireless or cabled Vantage Pro2 or Pro2 Plus. Half the cost of our 24-hour fan aspirated radiation shield, but nearly 75% as effective in reducing the effects of daytime radiation. Solar-powered fan runs during the day. Since there is no backup battery, the fan will cease running when night falls and the effects of radiation are less.
Kit includes fan, solar panel, additional radiation shield plates, and hardware.
1 Year Warranty
Specifications
  • DescriptionDay-time Fan Aspirated Radiation Shield Kit