Davis Instruments 7654 Solar-powered Long-range Repeater
Representative image only.
Davis Instruments 7654 Solar-powered Long-range Repeater
Davis Instruments | Mfr: 7654| Item: 3000341
Available for sale in the United States and Canada only.
$892.65CAD / Each
$892.65CAD / Each
Buy It Together
Current Item
Davis Instruments 7654 Solar-powered Long-range Repeater
Davis Instruments 7654 Solar-powered Long-range Repeater
3000341
$892.65CAD / Each
+ =
Total Price 1945.46 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with Vantage Pro2 and Vantage Vue only. Not compatible with original Vantage Pro.
  • 1 year warranty
More About this Item
Get the most from your Davis Instruments weather stations with these accessories and components.
This wireless long-range repeater with solar power is for those who wish to send weather data much farther than 1,000' (300 m). It can extend the range up to two miles (3.2 km).
Please note: Use of this product requires a pair of external antennas, sold separately. The long-range repeater has a transmitting and a receiving cable, each with an end which will connect to an external antenna. Please call to choose either our large omni directional antenna (#7656), range 1,560' (476 m) in all directions, or our Yagi directional antenna (#7660), range 5,000' (1.5 km) in one direction. You'll need two antennas for each long-range repeater. The charging system is capable of performing continuously even under low light conditions. Includes solar panel, regulator circuit, and rechargeable battery.
1 Year Warranty
Specifications
  • DescriptionSolar-powered Long-range Repeater