Cole-Parmer Two-Position Chilling/Heating Block, 100 VAC

Representative image only.

Cole-Parmer Two-Position Chilling/Heating Block, 100 VAC

Cole-Parmer | Item: 4417506

Chill, freeze, heat, or mix biological samples at your work station

$4,611.71CAD / Each

$4,611.71CAD / Each

Buy It Together

Current Item

Cole-Parmer Two-Position Chilling/Heating Block, 100 VAC
Cole-Parmer Two-Position Chilling/Heating Block, 100 VAC
4417506

$4,611.71CAD / Each
+ + + + =
Total Price 10514.84 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Chiller/heater module, power cord, instruction manual, and a 12 VDC power supply. Order cover and aluminum blocks separately.

Key Features

  • Store 5 programs in memory for precisely timed and repeatable routines
  • Choose blocks for centrifuge tubes, assay plates, PCR tubes & plates, and more
  • Two-block unit offers two different or same temperatures
  • 1 year warranty

More About this Item

Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM® compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100°C with ±1°C digital control.

Two-Position Chilling/Heating Block is non-programmable. It features two individually controlled blocks that can be set to the same temperature or two different temperatures.

1 Year WarrantyCanadian Standards AssociationUL Listed.
Specifications
  • Power (VAC)100 to 230
  • DescriptionTwo-Position Chilling/Heating Block, 100 VAC
  • Warranty1 year
Reviews
Q&A