Cole-Parmer ThinTouch™ Powder-Free Blue Disposable Nitrile Gloves, X-Large; 1000/CS
Representative image only.
Cole-Parmer ThinTouch™ Powder-Free Blue Disposable Nitrile Gloves, X-Large; 1000/CS
Cole-Parmer | Item: 1595771
Enhanced tactile sensitivity for a superior “touch & feel” experience
$128.92CAD / Case of 1000
BUY MORE & SAVE
  • 1-4$128.92 CAD / Case of 1000
  • 5+$116.03 CAD / Case of 1000
$128.92CAD / Case of 1000
BUY MORE & SAVE
  • 1-4$128.92 CAD / Case of 1000
  • 5+$116.03 CAD / Case of 1000
Buy It Together
Current Item
Cole-Parmer ThinTouch™  Powder-Free Blue Disposable Nitrile Gloves, X-Large; 1000/CS
Cole-Parmer ThinTouch™ Powder-Free Blue Disposable Nitrile Gloves, X-Large; 1000/CS
1595771
$128.92CAD / Case of 1000
+ + =
Total Price 258.73 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Advanced nitrile formulation with soft finish delivers optimal comfort and dexterity
  • Reliable protection against a wide range of chemicals
  • Textured fingertips ensure a secure grip in wet or dry conditions
  • High tensile strength for excellent tear and puncture resistance
  • Meets acceptable quality level (AQL) of 1.5
More About this Item
These nitrile gloves provide you protection against a variety of chemicals and are puncture resistant with a 3.2 mil palm thickness. The gloves offer a soft, comfortable fit, are latex- and powder-free, and help reduce fatigue. Textured fingertips enhance wet or dry grip. Available in 9.5” length and four sizes – small, medium, large, and X-large. Ambidextrous.
Conformite Europeenne
Specifications
  • Thickness (mil)3.2
  • MaterialNitrile
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffBeaded
  • ComplianceEN ISO 374-1:2016, AQL 1.5 (freedom from holes)
  • SterileNo
  • PowderedNo
  • DescriptionPowder-Free Blue Disposable Nitrile Gloves, X-Large; 1000/CS