Microflex DGP350-S Diamond Grip Plus Latex Gloves; Small
Representative image only.
Microflex DGP350-S Diamond Grip Plus Latex Gloves; Small
Microflex | Mfr: DGP350-S| Item: 8623230
Thick design provides durable protection
Call For Price
Call For Price
Buy It Together
Current Item
Microflex DGP350-S Diamond Grip Plus Latex Gloves; Small
Microflex DGP350-S Diamond Grip Plus Latex Gloves; Small
8623230
Call For Price
+ =
Total Price 15.77 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Medical examination grade protects hands of healthcare professionals
  • Fully textured surface offers secure grip
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
More About this Item
These exam gloves feature a palm thickness of 5.1 mil and a finger thickness of 6.3 mil for durable protection. Gloves feature a fully textured surface for excellent grip — wet or dry. Natural latex ensures superior comfort and resilience. Beaded cuffs improve tear resistance and protect hands.
Ideal for use in the food processing, life sciences, healthcare, utilities, and warehousing industries.
Standards and certifications include ASTM D6319, EN 420:2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
Please Note: This product contains natural rubber latex (latex) which may cause allergic reactions in some users. Safe use of this glove by or on latex sensitized individuals has not been established.
Specifications
  • MaterialLatex
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorClear
  • GripTextured
  • SterileNo
  • PowderedNo
  • Palm Thickness (mil)5.1
  • DescriptionDiamond Grip Plus Latex Gloves; size, small