Cole-Parmer Standard Manual Defrost Hazardous Location Freezer, Solid Door, -15°C to -25°C, 20 cubic ft, 120V

Representative image only.

Cole-Parmer Standard Manual Defrost Hazardous Location Freezer, Solid Door, -15°C to -25°C, 20 cubic ft, 120V

Cole-Parmer | Item: 4426247

Reliable cold storage for hazardous environments

$7,696.99CAD / Each
$9,621.24CAD / Each

$7,696.99CAD / Each
$9,621.24CAD / Each

Buy It Together

Current Item

Cole-Parmer Standard Manual Defrost Hazardous Location Freezer, Solid Door, -15°C to -25°C, 20 cubic ft, 120V
Cole-Parmer Standard Manual Defrost Hazardous Location Freezer, Solid Door, -15°C to -25°C, 20 cubic ft, 120V
4426247

$7,696.99CAD / Each
$9,621.24CAD / Each
+
Cole-Parmer Standard Refrigerator, Glass Door, 1°C to 10°C, 16 cubic ft, 120V
4426200

$5,221.70CAD / Each
$6,527.12CAD / Each
+ =
Total Price 20503.71 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Microprocessor temperature controller maintains stable conditions from -15°C to -25°C
  • Built for safety - Meets NFPA and OSHA guidelines for Hazardous Locations, Class 1, Division II, Group C & D
  • Advanced monitoring and security with digital display, audible/visual alarms, and remote alarm contacts for enhanced product protection
  • Environmentally friendly refrigerants, magnetic door gaskets, and cycle defrost for long-term reliability
  • 2 year warranty

More About this Item

Engineered for hazardous locations, these freezers offer precise temperature management and enhanced security features. A microprocessor temperature controller ensures stable low-temperature storage, while an advanced alarm system provides real-time monitoring for critical samples. Built to meet NFPA and OSHA standards, these units are designed for safe operation in volatile environments. With a rugged, hard-wired construction and eco-friendly refrigerants, they deliver reliable performance in demanding settings.

Two year warranty
Specifications
  • Freezer Capacity (cu ft)20
  • Min Temperature - Freezer (° F)-13
  • Max Temperature - Freezer (° F)5
  • Min Temperature - Freezer (° C)-25
  • Max Temperature - Freezer (° C)-15
  • Temperature ControlMicroprocessor
  • Freezer Interior Width (in)25 7/8
  • Freezer Interior Height (in)61 1/8
  • Freezer Interior Depth (in)21 3/4
  • Freezer Interior Width (cm)65.7225
  • Freezer Interior Height (cm)155.2575
  • Freezer Interior Depth (cm)55.245
  • Exterior Width (in)30 3/4
  • Exterior Height (in)74 5/8
  • Exterior Depth (in)29
  • Exterior Width (cm)78.105
  • Exterior Height (cm)189.5475
  • Exterior Depth (cm)73.66
  • Number of Doors1
  • Number of Shelves6
  • AlarmAudible and visual high and low temperature alarms
  • Cabinet ConstructionHigh density urethane foam cabinet construction
  • DisplayStandard Digital Temperature Display
  • Power (VAC)120
  • Power (Hz)60
  • Power (amps)5.5
  • DescriptionManual Defrost Hazardous Location Freezer, Solid Door, -15°C to -25°C, 20 cubic ft, 120V
  • Warranty2 years
Documents & Certificates
Reviews
Q&A