Cole-Parmer SHP-300-WS-120 Digital Stirring Hot Plate, 6" X 6", Aluminum, White; 120 VAC

Representative image only.

Cole-Parmer SHP-300-WS-120 Digital Stirring Hot Plate, 6" X 6", Aluminum, White; 120 VAC

Cole-Parmer | Item: 0480638

Digital stirring hot plates provide accurate stirring and temperature control

$1,354.05CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,354.05CAD / Each

Buy It Together

Current Item

Cole-Parmer SHP-300-WS-120 Digital Stirring Hot Plate, 6" X 6", Aluminum, White; 120 VAC
Cole-Parmer SHP-300-WS-120 Digital Stirring Hot Plate, 6" X 6", Aluminum, White; 120 VAC
0480638

$1,354.05CAD / Each
+ + + + + =
Total Price 5366.08 ADD 6 ITEMS TO CART
Product Description

Includes

Two 25-mm PTFE-coated stir bars and power cord with plug.

Key Features

  • Large 7-segment digital display shows set and actual temperature
  • ‘Hockey stick’ LED array displays speed
  • Hot warning light flashes when plate temperature is above 50 °C, even if unit is unplugged
  • Durable front panel resists spills and chemicals
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
  • Use optional digital temperature controller for precise sample temperature control
  • Ceramic-coated aluminum surface offers a robust top plate with excellent heat transmission
  • White body color
  • For 120 VAC operation
  • 3 year warranty
  • Previous part number SD152W/120

More About this Item

Ideal for day-to-day use by scientific professionals and students, the Cole-Parmer digital stirring hot plates offer precise stirring speed and temperature control of your samples. Powerful magnets provide consistent stirring speeds and the ability to mix large volumes up to 15 L.

The simple body design allows spillages to be easily deflected from the user. Front control panel resists spills and the glass is chemically toughened for extra resilience. In addition, units feature BioCote® antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.

The digital interface indicates the set and actual temperature on the 7-segment digital display. Stirring speed is displayed by a ‘hockey stick’ LED array with the lights progressively illuminating to represent the speed range.

To achieve accurate sample temperature control up to 200 °C (392 °F), a digital temperature controller (sold separately) can be connected to the rear of the unit.

The ceramic-coated aluminum plate offers excellent heat transmission, rapid heating with a 700-watt element, even temperature distribution, and is chemical resistant.

Applications: Reagent/media preparations, evaporating solvents, drying, boiling, warming, digestions, concentrations, extractions, synthesis

Please Note: BioCote® technology does not protect users or others against disease-causing bacteria, germs, viruses, or other harmful organisms. This technology is not a substitute for good hygiene and/or cleaning practices.

3 Year WarrantyConformite Europeenne
Specifications
  • Top Plate Length (in)6
  • Top Plate Width (in)6
  • Top Plate Length (cm)15
  • Top Plate Width (cm)15
  • Top Plate MaterialAluminum
  • Max Temperature (° F)617
  • Max Temperature (° C)325
  • Low Speed (rpm)50
  • High Speed (rpm)1400
  • Max Stirring Volume (Liters)15
  • Length (in)11 3/4
  • Width (in)7
  • Height (in)3 5/8
  • Length (cm)30
  • Width (cm)18
  • Height (cm)9.3
  • Power (VAC)120
  • Power (Hz)50/60
  • Power (watts)700
  • DescriptionSHP-300-WS-120 Digital Stirring Hot Plate, 6" X 6", Aluminum, White; 120 VAC
  • Warranty3 Years
  • ColorWhite
Reviews
Q&A