Cole-Parmer Melting Point Apparatus Calibration Key
Representative image only.
Cole-Parmer Melting Point Apparatus Calibration Key
Cole-Parmer | Item: 0301065
$637.24CAD / Each
$637.24CAD / Each
Buy It Together
Current Item
Product Description
Key Features
  • 1 year warranty
  • Previous part number AZ9253
More About this Item
Accessories compatible with digital melting point apparatus.
Use the calibration key along with certified calibration chemical to recalibrate your digital metling point apparatus.
1 Year Warranty
Specifications
  • Compatible WithMelting point apparatus 03010-15, 03010-41, 03010-51, 03010-59, and 03010-64
  • DescriptionMelting Point Apparatus Calibration Key