Cole-Parmer Light-duty Converter Clamp for 500- and 750-watt Units, Fits 1/2" dia rod
Representative image only.
Cole-Parmer Light-duty Converter Clamp for 500- and 750-watt Units, Fits 1/2" dia rod
Cole-Parmer | Item: 0471491
Enhance the performance of your ultrasonic processor
$127.89CAD / Each
$127.89CAD / Each
Buy It Together
Current Item
Cole-Parmer Light-duty Converter Clamp for 500- and 750-watt Units, Fits 1/2" dia rod
Cole-Parmer Light-duty Converter Clamp for 500- and 750-watt Units, Fits 1/2" dia rod
0471491
$127.89CAD / Each
+ + + + + + +
Cole-Parmer 500-Watt Ultrasonic Homogenizer, 115 VAC
0471170
$8,825.61CAD / Each
+ + =
Total Price 30459.46 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Compatible with Cole-Parmer 500- and 700-Watt ultrasonic processors
  • 3 year warranty
More About this Item
Choose from a wide range of accessories including support stands and converter clamps for your Cole-Parmer 500- and 700-Watt ultrasonic processors.
Light-duty converter clamp securely supports 2-1/2" (64 mm)diameter converters onto stands with 1/2" (13 mm) diameter support rod. Made of chemical-resistant reinforced plastic.
Recommended for light-duty applications.
For use with 500- and 750-watt ultrasonic processors. Order optional stand (04712-92) separately.
3 Year Warranty
Specifications
  • Compatible WithCole-Parmer 500 to 700 watt ultrasonic processors
  • Diameter (InchShort)1/2
  • DescriptionLight-duty Converter Clamp for 500- and 750-watt Units, Fits 1/2" dia rod