Cole-Parmer Light-duty Converter Clamp for 130-Watt Ultrasonic Processors, fits 1/2" dia rod
Representative image only.
Cole-Parmer Light-duty Converter Clamp for 130-Watt Ultrasonic Processors, fits 1/2" dia rod
Cole-Parmer | Item: 0471490
Enhance the performance of your ultrasonic processor
$127.89CAD / Each
$127.89CAD / Each
Buy It Together
Current Item
Cole-Parmer Light-duty Converter Clamp for 130-Watt Ultrasonic Processors, fits 1/2" dia rod
Cole-Parmer Light-duty Converter Clamp for 130-Watt Ultrasonic Processors, fits 1/2" dia rod
0471490
$127.89CAD / Each
+ + + + + + +
Cole-Parmer 500-Watt Ultrasonic Homogenizer, 115 VAC
0471170
$8,825.61CAD / Each
+ + =
Total Price 30459.46 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Compatible with Cole-Parmer 500- and 700-Watt ultrasonic processors
  • 3 year warranty
More About this Item
Choose from a wide range of accessories including support stands and converter clamps for your Cole-Parmer 500- and 700-Watt ultrasonic processors.
Clamp securely supports 1-1/4" (32 mm) dia converter onto stand with 1/2" (13 mm) dia support rod. Constructed of chemical resistant reinforced plastic.
Order support stand 04712-92 separately.
3 Year Warranty
Specifications
  • Compatible WithCole-Parmer 500 to 700 watt ultrasonic processors
  • DescriptionLight-duty Converter Clamp for 130-Watt Ultrasonic Processors, fits 1/2" dia rod