Cole-Parmer High-Accuracy Digital Gauge, 0 to 15 psia Absolute, 5-Digit LCD
Representative image only.
Cole-Parmer High-Accuracy Digital Gauge, 0 to 15 psia Absolute, 5-Digit LCD
Cole-Parmer | Item: 6893500
Durable and reliable digital pressure gauge
$518.63CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$518.63CAD / Each
Buy It Together
Current Item
Cole-Parmer High-Accuracy Digital Gauge, 0  to 15 psia Absolute, 5-Digit LCD
Cole-Parmer High-Accuracy Digital Gauge, 0 to 15 psia Absolute, 5-Digit LCD
6893500
$518.63CAD / Each
+ + + +
Cole-Parmer Digital Gauge, 760 Torr; 1/4"; NPT(M)
6893680
$623.98CAD / Each
+
SHOWA 1005-Md Latex Gloves, 5 mil, medium
8643151
$27.80CAD / Box of 100
$44.05CAD / Box of 100
+ +
Cole-Parmer Black Boot for Digital Pressure Gauges
6893695
$46.85CAD / Each
+
Cole-Parmer 40 Pound Nylon Cable Zip Ties, 8.0" L, Black; 1000/PK
0683088
$63.75CAD / Pkg of 1000
$95.62CAD / Pkg of 1000
+
Whatman 6713-0425 Venting Filter, Disc, 25mm, 50/pk
2970060
$518.26CAD / Pkg of 50
=
Total Price 4701.84 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Includes
2 AA batteries.
Key Features
  • High Accuracy with high repeatability
  • Backlilght and Zero-Reset Standard
  • Optional line powered with 4/20mA available & NPN limit switch
  • 1 year warranty
More About this Item
These digital pressure and vacuum gauges are an ideal alternative to your mechanical gauges! These gauges offer high accuracy plus the convenience of a digital display. The semiconductor sensing element measures pressure with ±0.25% or ±0.5% accuracy (depending on the range), and lasts for millions of cycles. These gauges come standard with Zero-Reset, Backlight, and Auto-Shutoff to help prolong battery life. Peak hold is standard on all pressure units.
As an option, some models can also come with a 4/20mA transmitter and NPN limit switch. These units are line powered with 8-30VDC.
Standard Models feature a
rugged, epoxy-coated extruded
aluminum case and rear cover
with ABS/polycarbonate front
bezel. Battery life up to 2000
hours; low-battery indication.
1 Year Warranty
Specifications
  • Accuracy±0.5% full-scale ±1 digit
  • Dial Size (in)3
  • Dial Size (mm)76.2
  • Max Pressure (psia)15
  • Wetted Materials316 SS
  • Case304 Stainless Steel
  • Process Connection1/4" NPT(M)
  • Unit Of Measurementpsi, kpa, bar, inH2O, inHg, Torr
  • Display5-digit LCD, 0.5" H
  • Battery Type2 AA (included)
  • Battery Life2000 hours
  • Media CompatibilityGases and liquids compatible with wetted materials
  • Resolution0.01 psi
  • DescriptionHigh-Accuracy Digital Gauge, 0 to 15 psia Absolute, 5-Digit LCD