SPAN Pressure Gauges for Petrochemical Applications
KemX® diaphragm prevents freezing and clogging
  • Accuracy of ±1% full-scale
  • Glycerin fill
  • High-resolution pressure intervals
These SPAN™ liquid-filled industrial pressure gauges feature the KemX diaphragm, located within the pressure connection, to prevent the freezing of the liquid fill while also preventing the clogging of the process connection. An additional feature of these gauges is the internal compensating diaphragm, eliminating internal case pressure that can cause shifts in accuracy during large changes in process and ambient temperature. The internal diaphragm also allows the liquid to be totally filled to the top of the gauge with no bubbles.
$102.84 - $250.84CAD / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Max Pressure (PSI)
Availability Pricing
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 15
$105.64CAD / Each
Thuemling LFS-210-30-G-KEMX (SG09008) 2.5" Filled Industrial Pressure Gauge, 0 to 30 psi 6808106 Mfr # LFS-210-30-G-KEMX (SG09008)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 30
$107.85CAD / Each
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 60
$107.85CAD / Each
Thuemling LFS-210-100-G-KEMX (SG09012) 2.5" Filled Industrial Pressure Gauge, 0 to 100 psi 6808110 Mfr # LFS-210-100-G-KEMX (SG09012)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 100
$102.99CAD / Each
Thuemling LFS-210-200-G-KEMX (SG09016) 2.5" Filled Industrial Pressure Gauge, 0 to 200 psi 6808114 Mfr # LFS-210-200-G-KEMX (SG09016)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 200
$106.67CAD / Each
Thuemling LFS-210-300-G-KEMX (SG09018) 2.5" Filled Industrial Pressure Gauge, 0 to 300 psi 6808116 Mfr # LFS-210-300-G-KEMX (SG09018)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 300
$102.84CAD / Each
Thuemling LFS-210-400-G-KEMX (SG09020) 2.5" Filled Industrial Pressure Gauge, 0 to 400 psi 6808118 Mfr # LFS-210-400-G-KEMX (SG09020)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 400
$102.84CAD / Each
Thuemling LFS-210-600-G-KEMX (SG09022) 2.5" Filled Industrial Pressure Gauge, 0 to 600 psi 6808120 Mfr # LFS-210-600-G-KEMX (SG09022)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 600
$102.84CAD / Each
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 1000
$102.84CAD / Each
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 3000
$102.84CAD / Each
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 2.5 5000
$102.84CAD / Each
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 30
$202.59CAD / Each
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 30
$199.64CAD / Each
Thuemling LFS-410-60-G-KEMX (SG09130) 4.5" Filled Industrial Pressure Gauge, 0 to 60 psi 6808138 Mfr # LFS-410-60-G-KEMX (SG09130)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 60
$212.90CAD / Each
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 100
$202.59CAD / Each
Thuemling LFS-410-160-G-KEMX (SG09134) 4.5" Filled Industrial Pressure Gauge, 0 to 160 psi 6808142 Mfr # LFS-410-160-G-KEMX (SG09134)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 160
$236.85CAD / Each
Thuemling LFS-410-200-G-KEMX (SG09136) 4.5" Filled Industrial Pressure Gauge, 0 to 200 psi 6808144 Mfr # LFS-410-200-G-KEMX (SG09136)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 200
$199.64CAD / Each
Thuemling LFS-410-2000-G-KEMX (SG09148) 4.5" Filled Industrial Pressure Gauge, 0 to 2000 psi 6808154 Mfr # LFS-410-2000-G-KEMX (SG09148)
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 2000
$204.43CAD / Each
COMPARE
±1% full-scale (ANSI/ASME Grade 1A) 4.5 5000
$250.84CAD / Each
Showing 1- 19 of 19