Cole-Parmer 10L Bag, TraceableOne™ Transport Bundle

Representative image only.

Cole-Parmer 10L Bag, TraceableOne™ Transport Bundle

Cole-Parmer | Item: 0372624

Confidently transport and monitor your temperature-sensitive samples

$1,382.04CAD / Each

$1,382.04CAD / Each
Product Description

Includes

Key Features

  • Improve sample protection – maintain specific temperature ranges for extended time periods
  • Choose from durable transport bag or box to meet your needs
  • Maintain sample integrity with Traceable® temperature monitoring
  • Can be used as outer packaging according to P650 per UN3373 (category B)*
  • Data logger provided with 2-year NIST-traceable calibration certificate
  • Modular cooling packs maintain 2°- 8°C for up to 15 hours
  • Lightweight insulated bags
  • 1 year warranty

More About this Item

The Cole-Parmer® transportation kit together with a Traceable® data logger is the perfect combination to safely transport your critical samples while maintaining sample integrity. Choose between a bag or box.

Transport Bag Kits feature an insulated nylon bag and modular cooling packs for flexible configurations to fit your samples. Bag has pockets to keep documentation safe during transit, a shoulder strap, and carrying handles. The bundle adds a pack of 10 TraceableOne™ single-use USB data loggers (model 18004-15). The logger is ready to use right out of the box – simply place it in your shipment and it will automatically generate a CSV report.

Transport Box Kits feature polypropylene-insulated walls for improved insulation and modular cooling packs for flexible configurations to fit your samples. The box has both a shoulder strap and carrying handles for easy transportation. The bundle adds a pack of 10 TraceableOne™ single-use USB data loggers (model 18004-15). The logger is ready to use right out of the box – simply place it in your shipment and it will automatically generate a CSV report.

*WARNING – product is suitable as outer packaging only according to P650 per UN3373 (category B).
To comply with shipping regulations for dangerous goods and infectious substances, additional packaging materials will be needed and packages containing infectious substances must be labelled in compliance with the U.S. Department of Transportation’s Hazardous Materials Regulations and the International Air Transport Association Dangerous Goods Regulations and other applicable laws and regulations. Failure to comply with these requirements may be violation of federal law.

The use of this product to transport dangerous goods or infectious substances should only be done by a certified dangerous goods specialist.

1 Year WarrantyNIST Traceable Calibration
Specifications
  • Volume (Liters)10
  • Lowest Set Temperature (° C)2
  • Capacity350 x 230 x 260 mm
  • Description10 L Bag, TraceableOne™ Transport Bundle
  • Warranty1 year
Reviews
Q&A