Chemetrics K-9400 Colorimetric Detergent Test Kit

Representative image only.

Chemetrics K-9400 Colorimetric Detergent Test Kit

Chemetrics | Mfr: K-9400| Item: 0554000

CHEMets test kits are portable and self-contained

$196.70CAD / Each

$196.70CAD / Each

Buy It Together

Current Item

Chemetrics K-9400 Colorimetric Detergent Test Kit
Chemetrics K-9400 Colorimetric Detergent Test Kit
0554000

$196.70CAD / Each
+ + + +
LaMotte Insta-Test Alkalinity Test Strips, 0 to 180 ppm
9953262

$43.02CAD / Pkg of 50
=
Total Price 605.26 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Vacuum sealed reagents and handy comparator chart for quick, visual results

More About this Item

CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.

The detergent test kit tests for anionic surfactants. The kit is provided in a cardboard box and does not include a polypropylene case.

Specifications
  • TestDetergents
  • Measurement Points0 to 3 ppm
  • Tests Per Kit20
  • DescriptionColorimetric Detergent Test Kit
Documents & Certificates
Reviews
Q&A