Chemetrics R-9400 Refill kit for 05540-00, 20/box (Detergent/anionic surfactant test refill)
Representative image only.
Chemetrics R-9400 Refill kit for 05540-00, 20/box (Detergent/anionic surfactant test refill)
Chemetrics | Mfr: R-9400| Item: 0554001
Take it wherever you go
$152.27CAD / Box of 20
$152.27CAD / Box of 20
Buy It Together
Current Item
Chemetrics R-9400 Refill kit for 05540-00, 20/box (Detergent/anionic surfactant test refill)
Chemetrics R-9400 Refill kit for 05540-00, 20/box (Detergent/anionic surfactant test refill)
0554001
$152.27CAD / Box of 20
+ + +
Chemetrics K-9010 Colorimetric Silica Test Kit
0554104
$152.27CAD / Each
=
Total Price 522.89 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed packaging keeps reagents active
More About this Item
CHEMets colorimetric test kits let you visually compare the color of your sample to standards using a comparator, comparator sold separately. Just break tip of ampoule in cup filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator provides accurate visual analysis.
Not Available for International Sales
Specifications
  • Compatible With05540-00
  • DescriptionRefill kit for 05540-00, 20/box (Detergent/anionic surfactant test refill)