Branson MC-2-QT Metal Caustic Free Alkaline Cleaner (emulsifying oils/greases/soils from Fe, SS, Ti, Cu), One Quart Bottle; 12/CS
Representative image only.
Branson MC-2-QT Metal Caustic Free Alkaline Cleaner (emulsifying oils/greases/soils from Fe, SS, Ti, Cu), One Quart Bottle; 12/CS
Branson | Mfr: MC-2-QT| Item: 0889816
Maximizes cleaning effectiveness of your ultrasonic cleaner
$353.25CAD / Case of 12
$353.25CAD / Case of 12
Buy It Together
Current Item
Branson MC-2-QT Metal Caustic Free Alkaline Cleaner (emulsifying oils/greases/soils from Fe, SS, Ti, Cu), One Quart Bottle; 12/CS
Branson MC-2-QT Metal Caustic Free Alkaline Cleaner (emulsifying oils/greases/soils from Fe, SS, Ti, Cu), One Quart Bottle; 12/CS
0889816
$353.25CAD / Case of 12
+ +
Cole-Parmer UC 350 Ultrasonic Cleaner, Heater/Mechanical Timer; 0.75 gal, 115V
0889539
$1,159.86CAD / Each
$1,442.45CAD / Each
+
Cole-Parmer UC 200 Ultrasonic Cleaner with Digital Timer and Heat, 20 Liter, 120 VAC
0889572
$1,196.40CAD / Each
$1,731.23CAD / Each
+ + + =
Total Price 21751.01 ADD 7 ITEMS TO CART
Product Description
Key Features
  • Ideal for use with all types of ultrasonic cleaners
  • Safely remove soils, rust, and oxides from a variety of materials
  • Specially formulated to remove oils, greases and a wide variety of solids from ferrous metals, stainless steel, titanium alloys, copper, and copper alloys
  • Biodegradable, phosphate and caustic free alkaline cleaner
More About this Item
Select the right cleaning solution to maximize the cleaning effectiveness of your ultrasonic cleaner. Cleaning solutions are formulated to deliver superior cleaning performance for a wide variety of cleaning applications.
Please Note: All the cleaning solutions are all biodegradable except for the liquid rust stripper.
Specifications
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)0.9
  • Dilution7-10% by volume, water
  • ApplicationsEmulsifying oils, greases, soils from Fe, SS, Ti, Cu
  • DescriptionMetal Caustic Free Alkaline Cleaner (emulsifying oils/greases/soils from Fe, SS, Ti, Cu), One Quart Bottle; 12/CS