Elma® Elmasonic xtra ST Industrial Ultrasonic Cleaners with Heat
Floor-standing units for high-throughput and industrial applications
  • Tackle the toughest of industrial cleaning needs with dual-frequency control
  • Select between sweep, pulse, and dynamic modes
  • Dependable and rugged—built for 24/7 operation
  • Easily preselect run time and temperature from the splashproof display
  • Move as needed with little effort—casters come standard
The Elmasonic xtra ST series of ultrasonic baths are constructed for rugged, continuous use. The stainless steel interior and exterior are built to withstand industrial wear and tear. The internal tank is sloped to drain thoroughly, and a side-mounted ball-valve allows for quick emptying of cleaning solutions. Operate at either 25 kHz for heavy parts and hard to remove soils, or at 45 kHz for more sensitive items.
Ultrasound comes from two generators in two locations—ideal for filter cleaning, even internal spaces can be reached. Operate in sweep, pulse, or dynamic modes. The sweep function ensures even sound distribution, pulse increases energy 20% for quick and effective treatment, and dynamic mode automatically switches between sweep and pulse modes to tackle even the toughest cleaning needs.
Select heating temperature from 30 to 80°C, in 5° increments, to help break down and remove oils. Units stand on industrial rollers to provide high mobility—perfect for production environments, workshops, and servicing. Optional lids help suppress noise production, and baskets allow for higher throughput. Choose from seven large sizes—all come with a 3-year warranty.
$16,126.25 - $38,786.99CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Frequency Output (KHz) Power (VAC)
Availability Pricing
Elmasonic xtra ST 600H Industrial Ultrasonic Cleaner with Heat, 15 gal, 240 VAC 5998880 Mfr # 107 7483
COMPARE
11.9 25 or 45 240
$16,126.25CAD / Each
Elmasonic xtra ST 800H Industrial Ultrasonic Cleaner with Heat, 22 gal, 200-208 VAC 5998881 Mfr # 107 7056
COMPARE
18.5 25 or 45 208
$21,349.42CAD / Each
Elmasonic xtra ST 1400H Industrial Ultrasonic Cleaner with Heat, 33 gal, 200-208 VAC 5998882 Mfr # 107 7058
COMPARE
25.6 25 or 45 208
$23,854.18CAD / Each
Elmasonic xtra ST 2500H Industrial Ultrasonic Cleaner with Heat, 67 gal, 200-208 VAC 5998884 Mfr # 107 7060
COMPARE
56.8 25 or 45 208
$38,786.99CAD / Each