Branson IS-QT Industrial Strength Caustic Free Alkaline Cleaner (heavy greases/oils/lubricants), One Quart Bottle; 12/CS
Representative image only.
Branson IS-QT Industrial Strength Caustic Free Alkaline Cleaner (heavy greases/oils/lubricants), One Quart Bottle; 12/CS
Branson | Mfr: IS-QT| Item: 0889814
Maximizes cleaning effectiveness of your ultrasonic cleaner
$351.40CAD / Case of 12
$351.40CAD / Case of 12
Buy It Together
Current Item
Branson IS-QT Industrial Strength Caustic Free Alkaline Cleaner (heavy greases/oils/lubricants), One Quart Bottle; 12/CS
Branson IS-QT Industrial Strength Caustic Free Alkaline Cleaner (heavy greases/oils/lubricants), One Quart Bottle; 12/CS
0889814
$351.40CAD / Case of 12
+ +
Cole-Parmer UC 350 Ultrasonic Cleaner, Heater/Mechanical Timer; 0.75 gal, 115V
0889539
$1,159.86CAD / Each
$1,442.45CAD / Each
+
Cole-Parmer UC 200 Ultrasonic Cleaner with Digital Timer and Heat, 20 Liter, 120 VAC
0889572
$1,196.40CAD / Each
$1,731.23CAD / Each
+ + + =
Total Price 21749.16 ADD 7 ITEMS TO CART
Product Description
Key Features
  • Ideal for use with all types of ultrasonic cleaners
  • Safely remove soils, rust, and oxides from a variety of materials
  • Specially formulated to remove heavy greases,oils, lubricants
  • Biodegradable, phosphate and caustic free alkaline cleaner
More About this Item
Select the right cleaning solution to maximize the cleaning effectiveness of your ultrasonic cleaner. Cleaning solutions are formulated to deliver superior cleaning performance for a wide variety of cleaning applications.
Please Note: All the cleaning solutions are all biodegradable except for the liquid rust stripper.
Specifications
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)0.9
  • Dilution8-10% by volume, water
  • ApplicationsHeavy greases, oils, lubricants
  • DescriptionIndustrial Strength Caustic Free Alkaline Cleaner, One Quart Bottle; 12/CS