Branson EC-QT Electronic Caustic Free Alkaline Cleaner (oils, resins, & rosins), One Quart Bottle; 12/CS

Representative image only.

Branson EC-QT Electronic Caustic Free Alkaline Cleaner (oils, resins, & rosins), One Quart Bottle; 12/CS

Branson | Mfr: EC-QT| Item: 0889813

Maximizes cleaning effectiveness of your ultrasonic cleaner

$688.07CAD / Case of 12

$688.07CAD / Case of 12

Buy It Together

Current Item

Branson EC-QT Electronic Caustic Free Alkaline Cleaner (oils, resins, & rosins), One Quart Bottle; 12/CS
Branson EC-QT Electronic Caustic Free Alkaline Cleaner (oils, resins, & rosins), One Quart Bottle; 12/CS
0889813

$688.07CAD / Case of 12
+
Cole-Parmer UC 200 Ultrasonic Cleaner with Digital Timer and Heat, 20 Liter, 120 VAC
0889572

$1,196.40CAD / Each
$1,731.23CAD / Each
+ + + + +
Cole-Parmer UC 350 Ultrasonic Cleaner, Heater/Mechanical Timer; 0.75 gal, 115V
0889539

$1,159.86CAD / Each
$1,442.45CAD / Each
=
Total Price 22085.83 ADD 7 ITEMS TO CART
Product Description

Key Features

  • Ideal for use with all types of ultrasonic cleaners
  • Safely remove soils, rust, and oxides from a variety of materials
  • Specially formulated to remove oils, resins, and rosins from hard surfaces
  • Contains no conductive metal cations
  • Biodegradable, phosphate and caustic free alkaline cleaner

More About this Item

Select the right cleaning solution to maximize the cleaning effectiveness of your ultrasonic cleaner. Cleaning solutions are formulated to deliver superior cleaning performance for a wide variety of cleaning applications.

Please Note: All the cleaning solutions are all biodegradable except for the liquid rust stripper.

Specifications
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)0.9
  • Dilution2-5% by volume, water
  • ApplicationsOils, resins, & rosins
  • DescriptionElectronic Caustic Free Alkaline Cleaner, One Quart Bottle; 12/CS
Reviews
Q&A