Branson BC-QT Buffing Compound Remover/Caustic Free Cleaner (tripoli, rouge, lime, diamond tripoli, etc), One Quart Bottle; 12/CS
Representative image only.
Branson BC-QT Buffing Compound Remover/Caustic Free Cleaner (tripoli, rouge, lime, diamond tripoli, etc), One Quart Bottle; 12/CS
Branson | Mfr: BC-QT| Item: 0889811
Maximizes cleaning effectiveness of your ultrasonic cleaner
$356.93CAD / Case of 12
$356.93CAD / Case of 12
Buy It Together
Current Item
Branson BC-QT Buffing Compound Remover/Caustic Free Cleaner (tripoli, rouge, lime, diamond tripoli, etc), One Quart Bottle; 12/CS
Branson BC-QT Buffing Compound Remover/Caustic Free Cleaner (tripoli, rouge, lime, diamond tripoli, etc), One Quart Bottle; 12/CS
0889811
$356.93CAD / Case of 12
+
Cole-Parmer UC 200 Ultrasonic Cleaner with Digital Timer and Heat, 20 Liter, 120 VAC
0889572
$1,196.40CAD / Each
$1,731.23CAD / Each
+ + + + +
Cole-Parmer UC 350 Ultrasonic Cleaner, Heater/Mechanical Timer; 0.75 gal, 115V
0889539
$1,159.86CAD / Each
$1,442.45CAD / Each
=
Total Price 21754.69 ADD 7 ITEMS TO CART
Product Description
Key Features
  • Ideal for use with all types of ultrasonic cleaners
  • Safely remove soils, rust, and oxides from a variety of materials
  • Specially formulated to remove tripoli, rouge, lime, diamond tripoli
  • Biodegradable, phosphate and caustic free cleaner
More About this Item
Select the right cleaning solution to maximize the cleaning effectiveness of your ultrasonic cleaner. Cleaning solutions are formulated to deliver superior cleaning performance for a wide variety of cleaning applications.
Please Note: All the cleaning solutions are all biodegradable except for the liquid rust stripper.
Specifications
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)0.9
  • Dilution6-7% by volume, water
  • ApplicationsTripoli, rouge, lime, diamond tripoli, etc
  • DescriptionBuffing Compound Remover/Caustic Free Cleaner, One Quart Bottle; 12/CS